Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: PRTDELKND LLKKYNVEYQ EYLQSKNKYK AEILKKLEHQ RLIEAERKRI AQMRQQQLES EQFLFFEDQL KKQELARGQM RSQQTSGLSE QIDGSALSCF STHQNNSLLN VFADQPNKSD ATNYASHSPP VNRALTPAAT LSAVQNLVVE GLRCVVLPED LCHKFLQLAE SNTVR (116-289 aa encoded by BC010846)
STAMBPL1 is zinc metalloprotease that specifically cleaves 'Lys-63'-linked polyubiquitin chains. Does not cleave 'Lys-48'-linked polyubiquitin chains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AMSH-like protease; AMSH-LP; associated molecule with the SH3 domain of STAM (AMSH) - Family Protein; associated molecule with the SH3 domain of STAM (AMSH) like protein; STAM binding protein-like 1; STAM-binding protein-like 1
Gene Aliases: ALMalpha; AMSH-FP; AMSH-LP; AMSHLP; bA399O19.2; KIAA1373; STAMBPL1
UniProt ID: (Human) Q96FJ0
Entrez Gene ID: (Human) 57559
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support