Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MMAKKPPKPA PRRIFQERLK ITALPLYFEG FLLIKRSGYR EYEHYWTELR GTTLFFYTDK KSIIYVDKLD IVDLTCLTEQ NSTEKNCAKF TLVLPKEEVQ LKTENTESGE EWRGFILTVT ELSVPQNVSL LPGQVIKLHE VLEREKKRRI ETEQSTSVEK EKEPTEDYVD VLNPMPACFY TVSRKEATEM LQKNPSLGNM ILRPGSDSRN YSITIRQEID IPRIKHYKVM SVGQNYTIEL EKPVTLPNLF SVIDYFVKET RGNLRPFICS TDENTGQEPS MEGRSEKLKK NPHIA
The protein encoded by this gene functions as a docking protein acting downstream of Tec tyrosine kinase in B cell antigen receptor signaling. The protein is directly phosphorylated by Tec in vitro where it participates in a postive feedback loop, increasing Tec activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BCR downstream signaling 1; BCR downstream-signaling protein 1; Docking protein BRDG1; Signal-transducing adaptor protein 1; STAP-1; Stem cell adaptor protein 1; stem cell adaptor protein STAP-1
Gene Aliases: AI586015; BRDG1; STAP-1; STAP1
UniProt ID: (Human) Q9ULZ2, (Mouse) Q9JM90
Entrez Gene ID: (Human) 26228, (Mouse) 56792
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support