Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Promo Code: RACCS25 Save on advanced culture solutions Save up to 22% on optimized formulations to streamline your neural, stem and cancer cell culture. Learn more
Immunogen sequence: MEKLAASTEP QGPRPVLGRE SVQVPDDQDF RSFRSECEAE VGWNLTYSRA GVSVWVQAVE MDRTLHKIKC RMECCDVPAE TLYDVLHDIE YRKKWDSNVI ETFDIARLTV NADVGYYSWR CPKPLKNRDV ITLRSWLPMG ADYIIMNYSV KHPKYPPRKD LVRAVSIQTG YLIQSTGPKS CVITYLAQVD PKGSLPKWVV NKSSQFLAPK AMKKMYKACL KYPEWKQKHL PHFKPWLHPE QSPLPSLALS ELSVQHADSL ENIDESAVAE SREERMGGAG GEGSDDDTSL T
STARD10 gene ontology annotations related to this gene include bile acid secretion; positive regulation of peroxisome proliferator activated receptor signaling pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antigen NY-CO-28; CGI52; PCTP-L; PCTP-like protein; serologically defined colon cancer antigen 10; Serologically defined colon cancer antigen 28; Serologically defined colon cancer antigen 28 homolog; StAR-related lipid transfer (START) domain containing 10; StAR-related lipid transfer domain containing 10; StAR-related lipid transfer protein 10; StARD10; START domain containing 10; START domain-containing protein 10
Gene Aliases: AV048538; CGI-52; NY-C0-28; NY-CO-28; PC-TP2; PCTP2; Pctpl; SDCCAG28; SdccagG28; STARD10; TISP-81
UniProt ID: (Human) Q9Y365, (Mouse) Q9JMD3
Entrez Gene ID: (Pig) 100511029, (Rabbit) 100337696, (Human) 10809, (Rat) 293150, (Mouse) 56018
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support