Product References
Aging-accelerated differential production and aggregation of STAT3 protein in neuronal cells and neural stem cells in the male mouse spinal cord.
Biogerontology
Zhao T,Liu C,Liu L,Wang X,Liu C
Wed Feb 01 00:00:00 EST 2023
Aging-accelerated differential production and aggregation of STAT3 protein in neuronal cells and neural stem cells in the male mouse spinal cord.
Biogerontology
Zhao T,Liu C,Liu L,Wang X,Liu C
Wed Feb 01 00:00:00 EST 2023
Aging-accelerated differential production and aggregation of STAT3 protein in neuronal cells and neural stem cells in the male mouse spinal cord.
Biogerontology
Zhao T,Liu C,Liu L,Wang X,Liu C
Wed Feb 01 00:00:00 EST 2023
Differences in p38-STAT3-S100A11 signaling after the administration of aristolochic acid I and IVa may account for the disparity in their nephrotoxicity.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Xian Z,Tian J,Zhao Y,Yi Y,Li C,Han J,Zhang Y,Wang Y,Wang L,Liu S,Pan C,Liu C,Wang D,Meng J,Tang X,Wang F,Liang A
Thu Jun 01 00:00:00 EDT 2023
4-Octyl itaconate attenuates LPS-induced acute kidney injury by activating Nrf2 and inhibiting STAT3 signaling.
Molecular medicine (Cambridge, Mass.)
Xu L,Cai J,Li C,Yang M,Duan T,Zhao Q,Xi Y,Sun L,He L,Tang C,Sun L
Mon Apr 24 00:00:00 EDT 2023
A novel GRK2 inhibitor alleviates experimental arthritis through restraining Th17 cell differentiation.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Tao J,Jiang C,Guo P,Chen H,Zhu Z,Su T,Zhou W,Tai Y,Han C,Ma Y,Chen J,Sun W,Wang Y,Wei W,Wang Q
Sun Jan 01 00:00:00 EST 2023
LC-MS Analysis of Ginsenosides in Different Parts of Panax quinquefolius and Their Potential for Coronary Disease Improvement.
Planta medica
Zhang X,Kong C,Wang X,Hou H,Yu H,Wang L,Li P,Li X,Zhang Y,Han L,Liu K
Thu Jun 01 00:00:00 EDT 2023
Pan-cancer analysis of PCAT6 and its effect on oesophageal squamous cell carcinoma cell proliferation and migration.
Medical oncology (Northwood, London, England)
Luo Q,Kuang Y,Tao X,Zhu X,Zhao X,Lu G,Zhang W,Sun E,Chen B
Wed Mar 22 00:00:00 EDT 2023
Nifuroxazide Activates the Parthanatos to Overcome TMPRSS2:ERG Fusion-Positive Prostate Cancer.
Molecular cancer therapeutics
Li C,Zhang J,Wu Q,Kumar A,Pan G,Kelvin DJ
Thu Mar 02 00:00:00 EST 2023
Sclareol attenuates liver fibrosis through SENP1-mediated VEGFR2 SUMOylation and inhibition of downstream STAT3 signaling.
Phytotherapy research : PTR
Ge MX,Niu WX,Bao YY,Lu ZN,He HW
Fri Sep 01 00:00:00 EDT 2023
A Bioinspired Manganese-Organic Framework Ameliorates Ischemic Stroke through its Intrinsic Nanozyme Activity and Upregulating Endogenous Antioxidant Enzymes.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Wang J,Wang Y,Xiaohalati X,Su Q,Liu J,Cai B,Yang W,Wang Z,Wang L
Sat Jul 01 00:00:00 EDT 2023
STAT3 promotes differentiation of monocytes to MDSCs via CD39/CD73-adenosine signal pathway in oral squamous cell carcinoma.
Cancer immunology, immunotherapy : CII
Cui H,Lan Z,Zou KL,Zhao YY,Yu GT
Mon May 01 00:00:00 EDT 2023
STAT3 promotes differentiation of monocytes to MDSCs via CD39/CD73-adenosine signal pathway in oral squamous cell carcinoma.
Cancer immunology, immunotherapy : CII
Cui H,Lan Z,Zou KL,Zhao YY,Yu GT
Mon May 01 00:00:00 EDT 2023
5'-tiRNA-Gln inhibits hepatocellular carcinoma progression by repressing translation through the interaction with eukaryotic initiation factor 4A-I.
Frontiers of medicine
Wu C,Liu D,Zhang L,Wang J,Ding Y,Sun Z,Wang W
Thu Jun 01 00:00:00 EDT 2023
Pi-Pa-Run-Fei-Tang alleviates lung injury by modulating IL-6/JAK2/STAT3/IL-17 and PI3K/AKT/NF-κB signaling pathway and balancing Th17 and Treg in murine model of OVA-induced asthma.
Journal of ethnopharmacology
Jie XL,Luo ZR,Yu J,Tong ZR,Li QQ,Wu JH,Tao Y,Feng PS,Lan JP,Wang P
Tue Dec 05 00:00:00 EST 2023
POR overexpression induces tamoxifen-resistance in breast cancer through the STAT1/c-Myc pathway.
Molecular carcinogenesis
Chen S,Wu D,Liu Q,Jin F,Yao F,Fang Y
Wed Feb 01 00:00:00 EST 2023
Rabeprazole destroyed gastric epithelial barrier function through FOXF1/STAT3-mediated ZO-1 expression.
Clinical and experimental pharmacology & physiology
Yang F,Li L,Zhou Y,Pan W,Liang X,Huang L,Huang J,Cheng Y,Geng L,Xu W,Gong S
Thu Jun 01 00:00:00 EDT 2023
Rabeprazole destroyed gastric epithelial barrier function through FOXF1/STAT3-mediated ZO-1 expression.
Clinical and experimental pharmacology & physiology
Yang F,Li L,Zhou Y,Pan W,Liang X,Huang L,Huang J,Cheng Y,Geng L,Xu W,Gong S
Thu Jun 01 00:00:00 EDT 2023
Rabeprazole destroyed gastric epithelial barrier function through FOXF1/STAT3-mediated ZO-1 expression.
Clinical and experimental pharmacology & physiology
Yang F,Li L,Zhou Y,Pan W,Liang X,Huang L,Huang J,Cheng Y,Geng L,Xu W,Gong S
Thu Jun 01 00:00:00 EDT 2023
Fetal anemia and elevated hepcidin in a mouse model of fetal alcohol spectrum disorder.
Pediatric research
Helfrich KK,Saini N,Kwan STC,Rivera OC,Mooney SM,Smith SM
Tue Aug 01 00:00:00 EDT 2023
Exosomal lncRNA HOTAIR induces PDL1+ B cells to impede anti-tumor immunity in colorectal cancer.
Biochemical and biophysical research communications
Xie Z,Xia J,Jiao M,Zhao P,Wang Z,Lin S,Xing Y,Li Y,Lu Z,Zhong Z,Miao C,Zhou P,Qian J,Wang L,Zhang D,Gu J,Chu Y,Liu R
Sun Feb 12 00:00:00 EST 2023
Exosomal lncRNA HOTAIR induces PDL1+ B cells to impede anti-tumor immunity in colorectal cancer.
Biochemical and biophysical research communications
Xie Z,Xia J,Jiao M,Zhao P,Wang Z,Lin S,Xing Y,Li Y,Lu Z,Zhong Z,Miao C,Zhou P,Qian J,Wang L,Zhang D,Gu J,Chu Y,Liu R
Sun Feb 12 00:00:00 EST 2023
Anemoside B4 protects against chronic relapsing colitis in mice by modulating inflammatory response, colonic transcriptome and the gut microbiota.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Han Q,Deng LR,Zou M,Tang HZ,Huang CY,Chen FJ,Tomlinson B,Li YH
Tue Nov 01 00:00:00 EDT 2022
YTHDF1 Negatively Regulates Treponema pallidum-Induced Inflammation in THP-1 Macrophages by Promoting SOCS3 Translation in an m6A-Dependent Manner.
Frontiers in immunology
Li Z,Teng M,Jiang Y,Zhang L,Luo X,Liao Y,Yang B
Fri Apr 22 00:00:00 EDT 2022
Afatinib Reverses EMT via Inhibiting CD44-Stat3 Axis to Promote Radiosensitivity in Nasopharyngeal Carcinoma.
Pharmaceuticals (Basel, Switzerland)
Huang H,Huang F,Liang X,Fu Y,Cheng Z,Huang Y,Chen Z,Duan Y,Chen Y
Tue Dec 27 00:00:00 EST 2022
Afatinib Reverses EMT via Inhibiting CD44-Stat3 Axis to Promote Radiosensitivity in Nasopharyngeal Carcinoma.
Pharmaceuticals (Basel, Switzerland)
Huang H,Huang F,Liang X,Fu Y,Cheng Z,Huang Y,Chen Z,Duan Y,Chen Y
Tue Dec 27 00:00:00 EST 2022
Afatinib Reverses EMT via Inhibiting CD44-Stat3 Axis to Promote Radiosensitivity in Nasopharyngeal Carcinoma.
Pharmaceuticals (Basel, Switzerland)
Huang H,Huang F,Liang X,Fu Y,Cheng Z,Huang Y,Chen Z,Duan Y,Chen Y
Tue Dec 27 00:00:00 EST 2022
Afatinib Reverses EMT via Inhibiting CD44-Stat3 Axis to Promote Radiosensitivity in Nasopharyngeal Carcinoma.
Pharmaceuticals (Basel, Switzerland)
Huang H,Huang F,Liang X,Fu Y,Cheng Z,Huang Y,Chen Z,Duan Y,Chen Y
Tue Dec 27 00:00:00 EST 2022
Protection of pancreatic β-cell by phosphocreatine through mitochondrial improvement via the regulation of dual AKT/IRS-1/GSK-3β and STAT3/Cyp-D signaling pathways.
Cell biology and toxicology
Wang H,Ai J,Shopit A,Niu M,Ahmed N,Tesfaldet T,Tang Z,Li X,Jamalat Y,Chu P,Peng J,Ma X,Qaed E,Han G,Zhang W,Wang J,Tang Z
Wed Jun 01 00:00:00 EDT 2022
KGF-2 Regulates STAP-2-Mediated Signal Transducer and Activator of Transcription 3 Signaling and Reduces Skin Scar Formation.
The Journal of investigative dermatology
Zhou Q,Gong J,Bi J,Yang X,Zhang L,Lu C,Li L,Chen M,Cai J,Yang R,Li X,Li Z,Wang X
Fri Jul 01 00:00:00 EDT 2022
KGF-2 Regulates STAP-2-Mediated Signal Transducer and Activator of Transcription 3 Signaling and Reduces Skin Scar Formation.
The Journal of investigative dermatology
Zhou Q,Gong J,Bi J,Yang X,Zhang L,Lu C,Li L,Chen M,Cai J,Yang R,Li X,Li Z,Wang X
Fri Jul 01 00:00:00 EDT 2022
Mucin 16 Promotes Colorectal Cancer Development and Progression Through Activation of Janus Kinase 2.
Digestive diseases and sciences
Liu Z,Gu Y,Li X,Zhou L,Cheng X,Jiang H,Huang Y,Zhang Y,Xu T,Yang W,Huang Q
Wed Jun 01 00:00:00 EDT 2022
Dihydromyricetin Alleviates Pulmonary Fibrosis by Regulating Abnormal Fibroblasts Through the STAT3/p-STAT3/GLUT1 Signaling Pathway.
Frontiers in pharmacology
Li Z,Geng J,Xie B,He J,Wang J,Peng L,Hu Y,Dai H,Wang C
Sat Apr 02 00:00:00 EDT 2022
Nicotinamide N-Methyltransferase Remodeled Cell Metabolism and Aggravated Proinflammatory Responses by Activating STAT3/IL1β/PGE2 Pathway.
ACS omega
Yang C,Wang T,Zhu S,Zong Z,Luo C,Zhao Y,Liu J,Li T,Liu X,Liu C,Deng H
Tue Oct 25 00:00:00 EDT 2022
Exposure to short-chain chlorinated paraffins induces astrocyte activation via JAK2/STAT3 signaling pathway.
Ecotoxicology and environmental safety
Ding W,Zhao Z,Zheng Y,Wang R,Zhang Z,Zhang Z,Wang X,Yu S,Liu L,Huang R,Zhao X,Wu Q
Thu Dec 15 00:00:00 EST 2022
Exposure to short-chain chlorinated paraffins induces astrocyte activation via JAK2/STAT3 signaling pathway.
Ecotoxicology and environmental safety
Ding W,Zhao Z,Zheng Y,Wang R,Zhang Z,Zhang Z,Wang X,Yu S,Liu L,Huang R,Zhao X,Wu Q
Thu Dec 15 00:00:00 EST 2022
Exposure to short-chain chlorinated paraffins induces astrocyte activation via JAK2/STAT3 signaling pathway.
Ecotoxicology and environmental safety
Ding W,Zhao Z,Zheng Y,Wang R,Zhang Z,Zhang Z,Wang X,Yu S,Liu L,Huang R,Zhao X,Wu Q
Thu Dec 15 00:00:00 EST 2022
Exposure to short-chain chlorinated paraffins induces astrocyte activation via JAK2/STAT3 signaling pathway.
Ecotoxicology and environmental safety
Ding W,Zhao Z,Zheng Y,Wang R,Zhang Z,Zhang Z,Wang X,Yu S,Liu L,Huang R,Zhao X,Wu Q
Thu Dec 15 00:00:00 EST 2022
Exposure to short-chain chlorinated paraffins induces astrocyte activation via JAK2/STAT3 signaling pathway.
Ecotoxicology and environmental safety
Ding W,Zhao Z,Zheng Y,Wang R,Zhang Z,Zhang Z,Wang X,Yu S,Liu L,Huang R,Zhao X,Wu Q
Thu Dec 15 00:00:00 EST 2022
Exposure to short-chain chlorinated paraffins induces astrocyte activation via JAK2/STAT3 signaling pathway.
Ecotoxicology and environmental safety
Ding W,Zhao Z,Zheng Y,Wang R,Zhang Z,Zhang Z,Wang X,Yu S,Liu L,Huang R,Zhao X,Wu Q
Thu Dec 15 00:00:00 EST 2022
Loss of RTN3 phenocopies chronic kidney disease and results in activation of the IGF2-JAK2 pathway in proximal tubular epithelial cells.
Experimental & molecular medicine
Fan LL,Du R,Liu JS,Jin JY,Wang CY,Dong Y,He WX,Yan RQ,Xiang R
Sun May 01 00:00:00 EDT 2022
Leptin activates the JAK/STAT pathway to promote angiogenesis in RF/6A cells in vitro.
International journal of ophthalmology
Zhang L,Li R,Wu BH,Liang TT,Liu Z,Ju W,Wang Y,Wen YT,Liu MC,Du JH
Sat Apr 23 00:00:00 EDT 2022
PRRX1 promotes colorectal cancer stemness and chemoresistance via the JAK2/STAT3 axis by targeting IL-6.
Journal of gastrointestinal oncology
Zhong L,Tan W,Yang Q,Zou Z,Zhou R,Huang Y,Qiu Z,Zheng K,Huang Z
Thu Dec 01 00:00:00 EST 2022
Transcription factor NFE2L3 promotes the proliferation of esophageal squamous cell carcinoma cells and causes radiotherapy resistance by regulating IL-6.
Computer methods and programs in biomedicine
Chen T,Xu B,Chen H,Sun Y,Song J,Sun X,Zhang X,Hua W
Tue Nov 01 00:00:00 EDT 2022
PRKAR2A-derived circular RNAs promote the malignant transformation of colitis and distinguish patients with colitis-associated colorectal cancer.
Clinical and translational medicine
Wan D,Wang S,Xu Z,Zan X,Liu F,Han Y,Jiang M,Wu A,Zhi Q
Tue Feb 01 00:00:00 EST 2022
Pathological Changes and Expression of JAK-STAT Signaling Pathway Hallmark Proteins in Rat Retinas at Different Time Points After Retinal Ischemia Reperfusion Injury.
Pathology oncology research : POR
Wang S,Yu A,Han M,Chen X,Li Z,Ke M,Cai X,Ai M,Xing Y
Mon May 09 00:00:00 EDT 2022
Identification of potential immunomodulators from Pulsatilla decoction that act on therapeutic targets for ulcerative colitis based on pharmacological activity, absorbed ingredients, and in-silico molecular docking.
Chinese medicine
Deng LR,Han Q,Zou M,Chen FJ,Huang CY,Zhong YM,Wu QY,Tomlinson B,Li YH
Thu Nov 24 00:00:00 EST 2022
Intestinal Flora Changes Induced by a High-Fat Diet Promote Activation of Primordial Follicles through Macrophage Infiltration and Inflammatory Factor Secretion in Mouse Ovaries.
International journal of molecular sciences
Fan Z,Zhang X,Shang Y,Zou M,Zhou M,E Q,Fei S,Chen W,Li J,Zhang X,Liu X
Wed Apr 27 00:00:00 EDT 2022
Comprehensive serum metabolomics and network analysis to reveal the mechanism of gypenosides in treating lung cancer and enhancing the pharmacological effects of cisplatin.
Frontiers in pharmacology
Qi YS,Xiao MY,Xie P,Xie JB,Guo M,Li FF,Piao XL
Thu Dec 22 00:00:00 EST 2022
Increased SPON1 promotes pancreatic ductal adenocarcinoma progression by enhancing IL-6 trans-signalling.
Cell proliferation
Huo Y,Yang J,Zheng J,Xu D,Yang M,Tao L,Yao H,Fu X,Yang J,Liu D,Hua R,Zhang J,Sun Y,Hu L,Liu W
Sun May 01 00:00:00 EDT 2022
Anemoside B4 ameliorates experimental autoimmune encephalomyelitis in mice by modulating inflammatory responses and the gut microbiota.
European journal of pharmacology
Zou M,Chen FJ,Deng LR,Han Q,Huang CY,Shen SS,Tomlinson B,Li YH
Thu Sep 15 00:00:00 EDT 2022
Propofol inhibited gastric cancer proliferation via the hsa-miR-328-3p/STAT3 pathway.
Clinical & translational oncology : official publication of the Federation of Spanish Oncology Societies and of the National Cancer Institute of Mexico
Bai ZM,Li XF,Yang Y,Yang YF,Lv DR,Tang LL
Wed Sep 01 00:00:00 EDT 2021
Selective STAT3 Inhibitor Alantolactone Ameliorates Osteoarthritis via Regulating Chondrocyte Autophagy and Cartilage Homeostasis.
Frontiers in pharmacology
Pei W,Huang X,Ni B,Zhang R,Niu G,You H
Sat Oct 16 00:00:00 EDT 2021
Dual Inhibition of Endoplasmic Reticulum Stress and Oxidation Stress Manipulates the Polarization of Macrophages under Hypoxia to Sensitize Immunotherapy.
ACS nano
Jiang M,Li X,Zhang J,Lu Y,Shi Y,Zhu C,Liu Y,Qin B,Luo Z,Du Y,Luo L,Peng L,You J
Tue Sep 28 00:00:00 EDT 2021
Dual Inhibition of Endoplasmic Reticulum Stress and Oxidation Stress Manipulates the Polarization of Macrophages under Hypoxia to Sensitize Immunotherapy.
ACS nano
Jiang M,Li X,Zhang J,Lu Y,Shi Y,Zhu C,Liu Y,Qin B,Luo Z,Du Y,Luo L,Peng L,You J
Tue Sep 28 00:00:00 EDT 2021
Switch receptor T3/28 improves long-term persistence and antitumor efficacy of CAR-T cells.
Journal for immunotherapy of cancer
Zhao S,Wang C,Lu P,Lou Y,Liu H,Wang T,Yang S,Bao Z,Han L,Liang X,Ma C,Gao L
Mon Nov 01 00:00:00 EDT 2021
Increased Nuclear Transporter KPNA2 Contributes to Tumor Immune Evasion by Enhancing PD-L1 Expression in PDAC.
Journal of immunology research
Zhou KX,Huang S,Hu LP,Zhang XL,Qin WT,Zhang YL,Yao LL,Yu Y,Zhou YQ,Zhu L,Ji J,Zhang ZG
Mon Nov 01 00:00:00 EDT 2021
Elevated Serum FGG Levels Prognosticate and Promote the Disease Progression in Prostate Cancer.
Frontiers in genetics
Peng HH,Wang JN,Xiao LF,Yan M,Chen SP,Wang L,Yang K
Tue May 18 00:00:00 EDT 2021
Uncovering the anti-NSCLC effects and mechanisms of gypenosides by metabolomics and network pharmacology analysis.
Journal of ethnopharmacology
Qi YS,Xie JB,Xie P,Duan Y,Ling YQ,Gu YL,Piao XL
Sun Dec 05 00:00:00 EST 2021
Noncanonical Wnt5a signaling regulates tendon stem/progenitor cells senescence.
Stem cell research & therapy
Chen M,Li Y,Xiao L,Dai G,Lu P,Rui Y
Mon Oct 18 00:00:00 EDT 2021
Combination therapy with ropivacaine-loaded liposomes and nutrient deprivation for simultaneous cancer therapy and cancer pain relief.
Theranostics
Zhang J,Zhu S,Tan Q,Cheng D,Dai Q,Yang Z,Zhang L,Li F,Zuo Y,Dai W,Chen L,Gu E,Xu G,Wei Z,Cao Y,Liu X
Thu May 13 00:00:00 EDT 2021
CCL5 derived from tumor-associated macrophages promotes prostate cancer stem cells and metastasis via activating β-catenin/STAT3 signaling.
Cell death & disease
Huang R,Wang S,Wang N,Zheng Y,Zhou J,Yang B,Wang X,Zhang J,Guo L,Wang S,Chen Z,Wang Z,Xiang S
Thu Apr 16 00:00:00 EDT 2020
CCL5 derived from tumor-associated macrophages promotes prostate cancer stem cells and metastasis via activating β-catenin/STAT3 signaling.
Cell death & disease
Huang R,Wang S,Wang N,Zheng Y,Zhou J,Yang B,Wang X,Zhang J,Guo L,Wang S,Chen Z,Wang Z,Xiang S
Thu Apr 16 00:00:00 EDT 2020
Inhibition of Connexin43 hemichannels with Gap19 protects cerebral ischemia/reperfusion injury via the JAK2/STAT3 pathway in mice.
Brain research bulletin
Chen B,Yang L,Chen J,Chen Y,Zhang L,Wang L,Li X,Li Y,Yu H
Fri Mar 01 00:00:00 EST 2019
Inhibition of Connexin43 hemichannels with Gap19 protects cerebral ischemia/reperfusion injury via the JAK2/STAT3 pathway in mice.
Brain research bulletin
Chen B,Yang L,Chen J,Chen Y,Zhang L,Wang L,Li X,Li Y,Yu H
Fri Mar 01 00:00:00 EST 2019
Exosome-transmitted p120-catenin suppresses hepatocellular carcinoma progression via STAT3 pathways.
Molecular carcinogenesis
Cheng Z,Lei Z,Yang P,Si A,Xiang D,Tang X,Guo G,Zhou J,Hüser N
Thu Aug 01 00:00:00 EDT 2019
Berberine Maintains the Neutrophil N1 Phenotype to Reverse Cancer Cell Resistance to Doxorubicin.
Frontiers in pharmacology
Zhang S,Zhou L,Zhang M,Wang Y,Wang M,Du J,Gu W,Kui F,Li J,Geng S,Du G
Fri Dec 22 00:00:00 EST 2023
Olean-28,13b-olide 2 plays a role in cisplatin-mediated apoptosis and reverses cisplatin resistance in human lung cancer through multiple signaling pathways.
Biochemical pharmacology
Zhu B,Ren C,Du K,Zhu H,Ai Y,Kang F,Luo Y,Liu W,Wang L,Xu Y,Jiang X,Zhang Y
Sun Dec 01 00:00:00 EST 2019
Alcohol's Dysregulation of Maternal-Fetal IL-6 and p-STAT3 Is a Function of Maternal Iron Status.
Alcoholism, clinical and experimental research
Saini N,Helfrich KK,Kwan STC,Huebner SM,Abazi J,Flentke GR,Blohowiak SE,Kling PJ,Smith SM
Fri Nov 01 00:00:00 EDT 2019
Postoperative resveratrol administration improves prognosis of rat orthotopic glioblastomas.
BMC cancer
Song X,Shu XH,Wu ML,Zheng X,Jia B,Kong QY,Liu J,Li H
Mon Sep 03 00:00:00 EDT 2018
Postoperative resveratrol administration improves prognosis of rat orthotopic glioblastomas.
BMC cancer
Song X,Shu XH,Wu ML,Zheng X,Jia B,Kong QY,Liu J,Li H
Mon Sep 03 00:00:00 EDT 2018
Postoperative resveratrol administration improves prognosis of rat orthotopic glioblastomas.
BMC cancer
Song X,Shu XH,Wu ML,Zheng X,Jia B,Kong QY,Liu J,Li H
Mon Sep 03 00:00:00 EDT 2018
Postoperative resveratrol administration improves prognosis of rat orthotopic glioblastomas.
BMC cancer
Song X,Shu XH,Wu ML,Zheng X,Jia B,Kong QY,Liu J,Li H
Mon Sep 03 00:00:00 EDT 2018
CEP55 Promotes Cell Motility via JAK2⁻STAT3⁻MMPs Cascade in Hepatocellular Carcinoma.
Cells
Li M,Gao J,Li D,Yin Y
Wed Aug 08 00:00:00 EDT 2018
CEP55 Promotes Cell Motility via JAK2⁻STAT3⁻MMPs Cascade in Hepatocellular Carcinoma.
Cells
Li M,Gao J,Li D,Yin Y
Wed Aug 08 00:00:00 EDT 2018
Effect of modified Xiaochaihu decoction‑containing serum on HepG2.2.15 cells via the JAK2/STAT3 signaling pathway.
Molecular medicine reports
Chen S,Wang Z,Wan S,Huang H,Liang H
Wed Nov 01 00:00:00 EDT 2017
Effect of modified Xiaochaihu decoction‑containing serum on HepG2.2.15 cells via the JAK2/STAT3 signaling pathway.
Molecular medicine reports
Chen S,Wang Z,Wan S,Huang H,Liang H
Wed Nov 01 00:00:00 EDT 2017
CD147 promotes Src-dependent activation of Rac1 signaling through STAT3/DOCK8 during the motility of hepatocellular carcinoma cells.
Oncotarget
Wang SJ,Cui HY,Liu YM,Zhao P,Zhang Y,Fu ZG,Chen ZN,Jiang JL
Thu Jan 01 00:00:00 EST 2015
CD147 promotes Src-dependent activation of Rac1 signaling through STAT3/DOCK8 during the motility of hepatocellular carcinoma cells.
Oncotarget
Wang SJ,Cui HY,Liu YM,Zhao P,Zhang Y,Fu ZG,Chen ZN,Jiang JL
Thu Jan 01 00:00:00 EST 2015