Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MCAERLGQFM TLALVLATFD PARGTDATNP PEGPQDRSSQ QKGRLSLQNT AEIQHCLVNA GDVGCGVFEC FENNSCEIRG LHGICMTFLH NAGKFDAQGK SFIKDALKCK AHALRHRFGC ISRKCPAIRE MVSQLQRECY LKHDLCAAAQ ENTRVIVEMI HFKDLLLHEP YVDLVNLLLT CGEEVKEAIT HSVQVQCEQN WGSLCSILSF CTSAIQKPPT APPERQPQVD RTKLSRAHHG EAGHHLPEPS SRETGRGAKG ERGSKSHPNA HARGRVGGLG AQGPSGSSEW EDEQSEYSDI RR
Has an anti-hypocalcemic action on calcium and phosphate homeostasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Stanniocalcin-2; Stanniocalcin-related protein; STC-2; STC-related protein
Gene Aliases: STC-2; STC2; STCRP
UniProt ID: (Human) O76061, (Rat) Q9R0K8
Entrez Gene ID: (Human) 8614, (Rat) 63878
Molecular Function: peptide hormone intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support