Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: EDLFQKILVP IQQVLKEGHL EKTEIDEVVL VGGSTRIPRI RQVIQEFFGK DPNTSVDPDL AVVTGVAIQA GIDGGSWPLQ VSALEIPNKH LQKTNF
STCH encodes a protein that is a member of the heat shock protein 70 family and is found associated with microsomes. Members of this protein family play a role in the processing of cytosolic and secretory proteins, as well as in the removal of denatured or incorrectly-folded proteins. The encoded protein contains an ATPase domain and has been shown to associate with a ubiquitin-like protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heat shock 70 kDa protein 13; heat shock protein 70kDa family member 13; heat shock protein 70kDa family, member 13; Heat shock protein family A member 13; microsomal stress 70 protein ATPase core; Microsomal stress-70 protein ATPase core; stress 70 protein chaperone microsome-associated 60 kDa protein; stress 70 protein chaperone, microsome-associated, 60kD; stress 70 protein chaperone, microsome-associated, 60kD human homolog; stress 70 protein chaperone, microsome-associated, 60kDa; stress 70 protein chaperone, microsome-associated, human homolog; Stress-70 protein chaperone microsome-associated 60 kDa protein; testis secretory sperm-binding protein Li 199a
Gene Aliases: 1600002I10Rik; AV006182; B230217N24Rik; HSPA13; STCH
UniProt ID: (Human) P48723, (Mouse) Q8BM72
Entrez Gene ID: (Human) 6782, (Mouse) 110920
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support