Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • STIM1 Antibodies

            Proteintech

            STIM1 Polyclonal Antibody

            78 References
            View all (34) STIM1 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite STIM1 Polyclonal Antibody

            • Antibody Testing Data (15)
            Application
            STIM1 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 15

            STIM1 Antibody (11565-1-AP) in ICC/IF

            Immunofluorescent analysis of HepG2 cells using 11565-1-AP (STIM1 antibody) at dilution of 1:25 and Rhodamine-Goat anti-Rabbit IGG.
            View Product

            STIM1 Antibody in Immunocytochemistry (ICC/IF)
            STIM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            STIM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            STIM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            STIM1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            STIM1 Antibody in Western Blot (WB)
            STIM1 Antibody in Western Blot (WB)
            STIM1 Antibody in Western Blot (WB)
            STIM1 Antibody in Western Blot (WB)
            STIM1 Antibody in Western Blot (WB)
            STIM1 Antibody in Western Blot (WB)
            STIM1 Antibody in Western Blot (WB)
            STIM1 Antibody in Western Blot (WB)
            STIM1 Antibody in Flow Cytometry (Flow)
            STIM1 Antibody in Immunoprecipitation (IP)

            Product Details

            11565-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:1,000
            View 43 publications 43 publications

            Immunohistochemistry (IHC)

            -
            View 8 publications 8 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:20-1:200
            -

            Immunocytochemistry (ICC/IF)

            1:10-1:100
            View 18 publications 18 publications

            Flow Cytometry (Flow)

            0.2 µg/1x10^6 cells
            View 3 publications 3 publications

            Immunoprecipitation (IP)

            0.5-4.0 µg
            View 3 publications 3 publications

            Miscellaneous PubMed (Misc)

            -
            View 1 publication 1 publication

            Not applicable (N/A)

            -
            View 2 publications 2 publications
            Product Specifications

            Species Reactivity

            Human, Mouse

            Published species

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            STIM1 Fusion Protein Ag2140 (2-350 aa encoded by BC021300)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.42 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            The MW of STIM1 is about 80-90 kDa which may be due to the phosphorylation or glycosylation forms. 11565-1-AP is rabbit polyclonal antibody raised against the N-terminal of human STIM1.

            Immunogen sequence: DVCVRLALW LLWGLLLHQG QSLSHSHSEK ATGTSSGANS EESTAAEFCR IDKPLCHSED EKLSFEAVRN IHKLMDDDAN GDVDVEESDE FLREDLNYHD PTVKHSTFHG EDKLISVEDL WKAWKSSEVY NWTVDEVVQW LITYVELPQY EETFRKLQLS GHAMPRLAVT NTTMTGTVLK MTDRSHRQKL QLKALDTVLF GPPLLTRHNH LKDFMLVVSI VIGVGGCWFA YIQNRYSKEH MKKMMKDLEG LHRAEQSLHD LQERLHKAQE EHRTVEVEKV HLEKKLRDEI NLAKQEAQRL KELREGTENE RSRQKYAEEE LEQVREALRK AEKELESHSS WYAPEALQKW (2-350 aa encoded by BC021300)

            Target Information

            STIM1 (stromal interacting molecule; also known as GOK) acts as a sensor of calcium depletion within the endoplasmic reticulum and transduces the signal to Orai1, the presumptive CRAC channel at the plasma membrane. Following decrease of luminal calcium concentration, STIM1 oligomerizes and induces Orai1 to enable entry of extracellular calcium into the cytoplasm. However, the precise mechanism of STIM1-Orai1 interaction has not been elucidated yet. Many questions also remain to be solved around STIM1 functional distribution. It turns out that STIM1 associates with growing ends of microtubules and is involved in endoplasmic reticulum tubule extension.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: STIM1L; Stromal interaction molecule 1

            View more View less

            Gene Aliases: D11S4896E; GOK; IMD10; SIM; STIM1; STRMK; TAM; TAM1

            View more View less

            UniProt ID: (Human) Q13586, (Mouse) P70302

            View more View less

            Entrez Gene ID: (Human) 6786, (Mouse) 20866

            View more View less

            Function(s)

            protease binding calcium channel regulator activity calcium ion binding protein binding identical protein binding microtubule plus-end binding store-operated calcium channel activity metal ion binding

            Process(es)

            store-operated calcium entry detection of calcium ion cellular calcium ion homeostasis activation of store-operated calcium channel activity regulation of calcium ion transport enamel mineralization regulation of cardiac conduction transport ion transport cation transport calcium ion transport myotube differentiation calcium ion transmembrane transport regulation of store-operated calcium entry
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-wg9fx:80/100.66.77.148:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0