Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MYAMKYMNKQ QCIERDEVRN VFRELEILQE IEHVFLVNLW YSFQDEEDMF MVVDLLLGGD LRYHLQQNVQ FSEDTVRLYI CEMALALDYL RGQHIIHRDV KPDNILLDER GHAHLTDFNI ATIIKDGERA TALAGTKPYM APEIFHSFVN GGTGYSFEVD WWSVGVMAYE LLRGWRPYDI HSSNAVESLV QLFSTVSVQY VPTWSKEMVA LLRKLLTVNP EHRLSSLQDV QAAPALAGVL WDHLSEKRVE PGFVPNKGRL HCDPTFELEE MILESRPLHK KKKRLAKNKS RDNSRDSSQS ENDYLQDCLD AIQQDFVIFN REKLKRSQDL PREPLPAPES RDAAEPVEDE AERSALPMCG PICPSAGSG
STK32C or YANK3 is a member of the YANK family and is a serine/threonine protein kinase of approximately 486 aa that requires magnesium for catalytic activity. There are two isoforms of the protein generated by alternative splicing and the protein undergoes serine phosphorylation by upstream protein kinases. The STK32C gene product is conserved in many species including chimpanzee, dog, cow, mouse, rat, and chicken.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PKE; PKE protein kinase; RP11-140A10.1; Serine/threonine-protein kinase 32C; stk32c; testicular tissue protein Li 187; Yet another novel kinase 3
Gene Aliases: MNCb-1563; PKE; Pkek; STK32C; YANK3
UniProt ID: (Human) Q5T0Q5, (Mouse) Q8QZV4
Entrez Gene ID: (Human) 282974, (Mouse) 57740
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support