Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MADSGLDKKS TKCPDCSSAS QKDVLCVCSS KTRVPPVLVV EMSQTSSIGS AESLISLERK KEKNINRDIT SRKDLPSRTS NVERKASQQQ WGRGNFTEGK VPHIRIENGA AIEEIYTFGR ILGKGSFGIV IEATDKETET KWAIKKVNKE KAGSSAVKLL EREVNILKSV KHEHIIHLEQ VFETPKKMYL VMELCEDGEL KEILDRKGHF SENETRWIIQ SLASAIAYLH NNDIVHRDLK LENIMVKSSL IDDNNEINLN IKVTDFGLAV KKQSRSEAML QATCGTPIYM APEVISAHDY SQQCDIWSIG VVMYMLLRGE PPFLASSEGK LFELIRKGEL HFENAVWNSI SDCAKSVLKQ LMKVDPAHRI TAKELLDNQW LTGNKLSSVR PTNVLEMMKE WKNNPESVEE NTTEEKNKPS TEEKLKSYQP WGNVPETNYT SDEEEEKQST TYEKQFPATS KDNFDMCSSS FTSSKLLPAE IKGEMEKTPV TPSQGTATKY PAKSGALSRT KKKL
STK33 is a serine/threonine kinase involved in signal transduction and cellular communication. STK33 gene ontology annotations related to this gene include ATP binding; protein kinase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Serine/threonine-protein kinase 33
Gene Aliases: STK33
UniProt ID: (Human) Q9BYT3
Entrez Gene ID: (Human) 65975
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support