Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MAMTAGTTTT FPMSNHTRER VTVAKLTLEN FYSNLILQHE ERETRQKKLE VAMEEEGLAD EEKKLRRSQH ARKETEFLRL KRTRLGLDDF ESLKVIGRGA FGEVRLVQKK DTGHIYAMKI LRKSDMLEKE QVAHIRAERD ILVEADGAWV VKMFYSFQDK RNLYLIMEFL PGGDMMTLLM KKDTLTEEET QFYISETVLA IDAIHQLGFI HRDIKPDNLL LDAKGHVKLS DFGLCTGLKK AHRTEFYRNL THNPPSDFSF QNMNSKRKAE TWKKNRRQLA YSTVGTPDYI APEVFMQTGY NKLCDWWSLG VIMYEMLIGY PPFCSETPQE TYRKVMNWKE TLVFPPEVPI SEKAKDLILR FCIDSENRIG NSGVEEIKGH PFFEGVDWEH IRERPAAIPI EIKSIDDTSN FDDFPESDIL QPVPNTTEPD YKSKDWVFLN YTYKRFEGLT QRGSIPTYMK AGKL
STK38L (also known as NDR2) or Nuclear Dbf2-related protein is a serine/threonine kinase in the subgroup of the AGC kinase family. STK38L act as novel pro-apoptotic kinase that is activated by RASSF1A/MST1 in response to Fas receptor stimulation and promotes apoptosis. STK38L is mainly involved in the regulation of structural processes in differentiating and mature neuronal cells and it is highly expressed with highest levels observed in the thymus. STK38L contains 12 protein kinase catalytic subdomains and a potential bipartite nuclear localization signal.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NDR2 protein kinase; nuclear Dbf2-related 2; Nuclear Dbf2-related kinase 2; Serine/threonine-protein kinase 38-like; STK38L
Gene Aliases: KIAA0965; NDR2; STK38L
UniProt ID: (Human) Q9Y2H1
Entrez Gene ID: (Human) 23012
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support