Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Peptide sequence: MEGAGAGSGF RKELVSRLLH LHFKDDKTKE AAVRGVRQAQ AEDALRVDVD
Sequence homology: Human: 100%
STRA13 is a DNA-binding component of the FA core complex involved in DNA damage repair and genome maintenance.STRA13 is recruited to forks stalled by DNA interstrand cross-links, and required for cellular resistance to such lesions. As a component of the APITD1/CENPS complex, STRA13 is also essential for the stable assembly of the outer kinetchore.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CENP-X; Centromere protein X; FANCM-associated histone fold protein 2; FANCM-interacting histone fold protein 2; Fanconi anemia-associated polypeptide of 10 kDa; MGC14480; Retinoic acid-inducible gene D9 protein homolog; stimulated by retinoic acid 13 homolog; Stimulated by retinoic acid gene 13 protein homolog
Gene Aliases: CENP-X; CENPX; D9; FAAP10; MHF2; STRA13
UniProt ID: (Human) A8MT69
Entrez Gene ID: (Human) 201254
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support