Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSCRDRTQEF LSACKSLQTR QNGIQTNKPA LRAVRQRSEF TLMAKRIGKD LSNTFAKLEK LTILAKRKSL FDDKAVEIEE LTYIIKQDIN SLNKQIAQLQ
Mediates endoplasmic reticulum to Golgi transport. Together with p115/USO1 and GM130/GOLGA2, involved in vesicle tethering and fusion at the cis-Golgi membrane to maintain the stacked and inter-connected structure of the Golgi apparatus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: syntaxin 5A; Syntaxin-5
Gene Aliases: SED5; STX5; STX5A
UniProt ID: (Human) Q13190, (Rat) Q08851
Entrez Gene ID: (Human) 6811, (Rat) 65134
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support