Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSYTPGVGGD PAQLAQRISS NIQKITQCSV EIQRTLNQLG TPQDSPELRQ QLQQKQQYTN QLAKETDKYI KEFGSLPTTP SEQRQRKIQK DRLVAEFTTS LTNFQKVQRQ AAEREKEFVA RVRASSRVSG SFPEDSSKER NLVSWESQTQ PQVQVQDEEI TEDDLRLIHE RESSIRQLEA DIMDINEIFK DLGMMIHEQG DVIDSIEANV ENAEVHVQQA NQQLSRAADY QRKSRKTLCI IILILVIGVA IISLIIWGLN H
ZCRB1 is a nuclear protein first identified in a differential display screen involving morphine-dependence-related genes. Its expression is increased following morphine treatment and has been observed to be elevated in HepG2 cells. ZCRB1 contains a CCHC-type zing finger RNA-binding motif and can interact with the DBA-binding domain of the stem cell regulator C/EBP transcription factors. ZCRB1 was identified as one of the protein components of U11/U12 snRNPs, which are components of U12-type spliceosome and function as a molecular bridge connecting both ends of the intron, suggesting ZCRB1 may play a key role in U12-type splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Syntaxin-7; Syntaxin7
Gene Aliases: STX7
UniProt ID: (Human) O15400
Entrez Gene ID: (Human) 8417
Molecular Function: SNARE protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support