Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MAPDPWFSTY DSTCQIAQEI AEKIQQRNQY ERKGEKAPKL TVTIRALLQN LKEKIALLKD LLLRAVSTHQ ITQLEGDRRQ NLLDDLVTRE RLLLASFKNE GAEPDLIRSS LMSEEAKRGA PNPWLFEEPE ETRGLGFDEI RQQQQKIIQE QDAGLDALSS IISRQKQMGQ EIGNELDEQN EIIDDLANLV ENTDEKLRNE TRRVNMVDRK SASCGMIMVI LLLLVAIVVV AVWPTN
The gene is a member of the syntaxin family. The encoded protein is involved in protein trafficking from early to late endosomes via vesicle fusion and exocytosis. A related pseudogene has been identified on chromosome 12. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CIP-1-associated regulator of cyclin B; Syntaxin-8
Gene Aliases: CARB; STX8
UniProt ID: (Human) Q9UNK0
Entrez Gene ID: (Human) 9482
Molecular Function: SNARE protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support