Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous with the corresponding sequence in rat, mouse, chicken and monkey.
Specificity of this antibody: Syntaxin-binding protein 1.
This gene encodes a syntaxin-binding protein. The encoded protein appears to play a role in release of neurotransmitters via regulation of syntaxin, a transmembrane attachment protein receptor. Mutations in this gene have been associated with infantile epileptic encephalopathy-4. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EIEE4; FLJ37475; hUNC18; MGC146482; minisatellite 10g detected by probe MMS10; munc-18c; MUNC18-1; N-Sec1; neuronal SEC1; p67; Protein unc-18 homolog 1; Protein unc-18 homolog A; rbSec1; rop; RP11-56D16.3; Syntaxin-binding protein 1; UNC-18 homolog; unc-18-1; Unc-18A; Unc18-1
Gene Aliases: AI317162; AI326233; ANC18HA; MMS10-G; Ms10g; Munc-18a; MUNC18-1; N-sec1; NSEC1; NSEC1A; P67; Rb-sec1; RBSEC1; rbSec1A; rbSec1B; Sec1; STXBP1; Sxtbp1; UNC18; Unc18-1; UNC18A; Unc18h
UniProt ID: (Human) P61764, (Mouse) O08599, (Rat) P61765
Entrez Gene ID: (Human) 6812, (Mouse) 20910, (Rat) 25558
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support