Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: HELTFQAMS YDLLPIENDV YKYETSGIGE ARVKEVLLDE DDDLWIALRH KHIAEVSQEV TRSLKDFSSS KRMNTGEKTT MRDLSQMLKK MPQYQKELSK YSTHLHLAED CMKHYQGTVD KLCRVEQDLA MGTDAEGEKI KDPMRAIVPI LLDANVSTYD KIRIILLYIF LKNGITEENL NKLIQHAQIP PEDSEIITNM AHLGVPIVTD STLRRRSKPE RKERISEQTY QLSRWTPIIK DIMEDTIEDK LDTKHYPYIS TRSSASFSTT AVSARYGHWH KNKAPGEYRS GPRLIIFILG GVSLNEMRCA YEVTQANGKW EVLIGSTHIL TPQKLLDTLK KLNKTDEEIS S (245-594 aa encoded by BC015749)
This gene encodes a syntaxin-binding protein. The encoded protein appears to play a role in release of neurotransmitters via regulation of syntaxin, a transmembrane attachment protein receptor. Mutations in this gene have been associated with infantile epileptic encephalopathy-4. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EIEE4; FLJ37475; hUNC18; MGC146482; minisatellite 10g detected by probe MMS10; munc-18c; MUNC18-1; N-Sec1; neuronal SEC1; p67; Protein unc-18 homolog 1; Protein unc-18 homolog A; rbSec1; rop; RP11-56D16.3; Syntaxin-binding protein 1; UNC-18 homolog; unc-18-1; Unc-18A; Unc18-1
Gene Aliases: AI317162; AI326233; ANC18HA; MMS10-G; Ms10g; Munc-18a; MUNC18-1; N-sec1; NSEC1; NSEC1A; P67; Rb-sec1; RBSEC1; rbSec1A; rbSec1B; Sec1; STXBP1; Sxtbp1; UNC18; Unc18-1; UNC18A; Unc18h
UniProt ID: (Human) P61764, (Mouse) O08599, (Rat) P61765
Entrez Gene ID: (Human) 6812, (Mouse) 20910, (Rat) 25558
Molecular Function: membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support