Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: REQRTQQQRS GPQGIAPVPP PRDLSWEAGH GGNVALPLKE TSVENFLGAT TPALAKLQVP REQLSEVLEQ ICSGSCGPIF RANMNTGDPS KPKSVILKAL KEPAGLHEVQ
STYK1, a probable tyrosine protein-kinase, which has strong transforming capabilities on a variety of cell lines. When overexpressed, it can also induce tumor cell invasion as well as metastasis in distant organs. May act by activating both MAP kinase and phosphatidylinositol 3'-kinases (PI3K) pathways. It is widely expressed; highly expressed in brain, placenta and prostate. STYK1 is expressed in tumor cells such as hepatoma cells LO2, cervix carcinoma cells HeLa, ovary cancer cells Ho8910 and chronic myelogenous leukemia cells K562, but not in other tumor cells such as epidermoid carcinoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp761P1010; Novel oncogene with kinase domain; Protein PK-unique; Serine/threonine/tyrosine kinase 1; Tyrosine-protein kinase STYK1
Gene Aliases: NOK; STYK1; SuRTK106
UniProt ID: (Human) Q6J9G0
Entrez Gene ID: (Human) 55359
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support