Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MVSGSSGLA AARLLSRSFL LPQNGIRHCS YTASRQHLYV DKNTKIICQG FTGKQGTFHS QQALEYGTKL VGGTTPGKGG QTHLGLPVFN TVKEAKEQTG ATASVIYVPP PFAAAAINEA IEAEIPLVVC ITEGIPQQDM VRVKHKLLRQ EKTRLIGPNC PGVINPGECK IGIMPGHIHK KGRIGIVSRS GTLTYEAVHQ TTQVGLGQSL CVGIGGDPFN GTDFIDCLEI FLNDSATEGI ILIGEIGGNA EENAAEFLKQ HNSGPNSKPV VSFIAGLTAP PGRRMGHAGA IIAGGKGGAK EKISALQSAG VVVSMSPAQL GTTIYKEFEK RKML (1-333 aa encoded by BC000504)
This gene encodes the alpha subunit of the heterodimeric enzyme succinate coenzyme A ligase. This enzyme is targeted to the mitochondria and catalyzes the conversion of succinyl CoA and ADP or GDP to succinate and ATP or GTP. Mutations in this gene are the cause of the metabolic disorder fatal infantile lactic acidosis and mitochondrial DNA depletion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SCS-alpha; Succinate--CoA ligase [ADP/GDP-forming] subunit alpha, mitochondrial; succinate-CoA ligase, alpha subunit; succinate-CoA ligase, GDP-forming alpha subunit; succinyl-CoA ligase [ADP/GDP-forming] subunit alpha, mitochondrial; succinyl-CoA ligase [GDP-forming] subunit alpha, mitochondrial; succinyl-CoA synthetase alpha subunit; Succinyl-CoA synthetase subunit alpha
Gene Aliases: 1500000I01Rik; GALPHA; MTDPS9; SUCLA1; SUCLG1
UniProt ID: (Human) P53597, (Rat) P13086, (Mouse) Q9WUM5
Entrez Gene ID: (Human) 8802, (Rat) 114597, (Mouse) 56451
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support