Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive Control: Human kidney tissue lysates, rat kidney tissue lysates, human small intestine tissue.
Subcellular Location: Cytosol.
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes a protein that transfers a sulfo moiety to and from estrone, which may control levels of estrogen receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EST-1; EST-2; EST-6; Estrogen sulfotransferase; Estrogen sulfotransferase, isoform 1; Estrogen sulfotransferase, isoform 6; Estrone sulfotransferase; ST1E1; Sulfotransferase 1E1; sulfotransferase family 1E, estrogen-preferring, member 1; Sulfotransferase, estrogen-preferring
Gene Aliases: EST; EST-1; ST1E1; STE; Ste1; Ste2; SULT1E1
UniProt ID: (Human) P49888, (Rat) P52844
Entrez Gene ID: (Human) 6783, (Rat) 360268
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support