Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MANEKPTEEV KTENNNHINL KVAGQDGSVV QFKIKRQTPL SKLMKAYCEP RGLSVKQIRF RFGGQPISGT DKPAQLEMED EDTIDVFQQP TGGVY
SUMO4 is a member of the SUMO gene family. This family of small ubiquitin-related modifiers covalently modify target lysines in proteins and control the target proteins' subcellular localization, stability, or activity. Upon oxidative stress, SUMO4 conjugates to various anti-oxidant enzymes, chaperones, and stress defense proteins. This protein may also conjugate to NFKBIA, TFAP2A and FOS, negatively regulating their transcriptional activity, and to NR3C1, positively regulating its transcriptional activity. Covalent attachment to SUMO4 substrates requires prior activation by the E1 complex SAE1-SAE2 and linkage to the E2 enzyme UBE2I. In contrast to SUMO1, SUMO2 and SUMO3, SUMO4 seems to be insensitive to sentrin-specific proteases due to the presence of Pro-90. This may impair processing to mature form and conjugation to substrates. SUMO4 is located in the cytoplasm and specifically modifies IKBA, leading to negative regulation of NF-kappa-B-dependent transcription of the IL12B gene. The M55V substitution has been associated with type I diabetes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: insulin-dependent diabetes mellitus 5; small ubiquitin-like modifier 4 protein; Small ubiquitin-like protein 4; Small ubiquitin-related modifier 4; SMT3 suppressor of mif two 3 homolog 2; SMT3 suppressor of mif two 3 homolog 4; SUMO 4; SUMO-4
Gene Aliases: dJ281H8.4; IDDM5; SMT3H4; SUMO-4; SUMO4
UniProt ID: (Human) Q6EEV6
Entrez Gene ID: (Human) 387082
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support