Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YPYGLQTFH PALVSWWAAK DSRRPDEGWE ARDSSPHFQA EQRVMSRVHS LERRLEALAA EFSSNWQKEA MRLERLELRQ GAPGQGGGGG LSHEDTLALL EGLVSRREAA LKEDFRRETA ARIQEELSAL RAEHQQDSED LFKKIVRASQ ESEARIQQLK SEWQSMTQES FQESSVKELR RLEDQLAGLQ QELAALALKQ SSVAEEVGLL PQQIQAVRDD VESQFPAWIS QFLARGGGGR VGLLQREEMQ AQLRELESKI LTHVAEMQGK SAREAAASLS LTLQKEGVIG VTEEQVHHIV KQALQRYSED RIGLADYAL (234-551 aa encoded by BC111717)
UNC84B may form a physical interaction between the nuclear envelope and the centrosome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: nuclear envelope protein; Protein unc-84 homolog B; Rab5-interacting protein; Rab5IP; Sad1 unc-84 domain protein 2; Sad1/unc-84 protein-like 2; SUN domain-containing protein 2; unc-84 homolog B
Gene Aliases: B230369L08Rik; C030011B15; FRIGG; KIAA0668; RAB5IP; SUN2; UNC84B
UniProt ID: (Human) Q9UH99, (Mouse) Q8BJS4
Entrez Gene ID: (Human) 25777, (Mouse) 223697
Molecular Function:
cytoskeletal protein
microtubule or microtubule-binding cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support