Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: APINKKQPFV CSSLLQFARQ YSRNEPLTFA WLRRYIKWPL LPPKNIKDLM DLEAVHDVLD LYLWLSYRFM DMFPDASLIR DLQKELDGII QDGVHNITKL IKMSETHKLL NLEGFPSGSQ SRLSGTLKSQ ARRTRGTKAL GSKATEPPSP DAGELSLASR LVQQGLLTPD MLKQLEKEWM TQQTEHNKEK TESGTHPKGT RRKKKEPDSD; Positive Samples: K-562, Raji, Jurkat, SW620, Mouse heart, Mouse kidney, Mouse liver; Cellular Location: Mitochondrion matrix, Nucleus, mitochondrion nucleoid
Major helicase player in mitochondrial RNA metabolism. Component of the mitochondrial degradosome (mtEXO) complex, that degrades 3' overhang double-stranded RNA with a 3'-to-5' directionality in an ATP-dependent manner. Involved in the degradation of non-coding mitochondrial transcripts (MT-ncRNA) and tRNA-like molecules (PubMed:29967381). ATPase and ATP-dependent multisubstrate helicase, able to unwind double-stranded (ds) DNA and RNA, and RNA/DNA heteroduplexes in the 5'-to-3' direction. Plays a role in the RNA surveillance system in mitochondria; regulates the stability of mature mRNAs, the removal of aberrantly formed mRNAs and the rapid degradation of non coding processing intermediates. Also implicated in recombination and chromatin maintenance pathways. May protect cells from apoptosis. Associates with mitochondrial DNA. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent RNA helicase SUPV3L1, mitochondrial; Suppressor of var1 3-like protein 1; suppressor of var1, 3-like 1; suppressor of var1, 3-like 1(SUV3); SUV3-like helicase; SUV3-like protein 1
Gene Aliases: 6330443E10Rik; SUPV3L1; SUV3; Suv3l1
UniProt ID: (Human) Q8IYB8, (Mouse) Q80YD1, (Rat) Q5EBA1
Entrez Gene ID: (Human) 6832, (Mouse) 338359, (Rat) 294385
Molecular Function:
DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support