Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MAQQRALPQS KETLLQSYNK RLKDDIKSIM DNFTEIIKTA KIEDETQVSR ATQGEQDNYE MHVRAANIVR AGESLMKLVS DLKQFLILND FPSVNEAIDQ
This gene is located in the surfeit gene cluster, a group of very tightly linked housekeeping genes that do not share sequence similarity. The gene is oriented in a head-to-head fashion with RPL7A (SURF3) and the two genes share a bidirectional promoter. The encoded proteins are localized to the cytoplasm. Two alternative transcript variants encoding different isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mediator complex subunit 22; Mediator of RNA polymerase II transcription subunit 22; MGC48682; Surf-5; Surfeit locus protein 5
Gene Aliases: MED22; MED24; surf-5; SURF5
UniProt ID: (Human) Q15528
Entrez Gene ID: (Human) 6837
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support