Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Probably plays a role in the control of regulated secretion in neural and endocrine cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Synaptic vesicle glycoprotein 2B; Synaptic vesicle protein 2B; synaptic vesicle protein 2B homolog
Gene Aliases: A830038F04Rik; AI850768; HsT19680; KIAA0735; mKIAA0735; SV2B
UniProt ID: (Human) Q7L1I2, (Mouse) Q8BG39
Entrez Gene ID: (Human) 9899, (Mouse) 64176
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support