Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Peptide Sequence: YFQDEEYKSK MKVFFGEHVY GATINFTMEN QIHQHGKLVN DKFTRMYFKH VLFEDTFFDE
Probably plays a role in the control of regulated secretion in neural and endocrine cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Synaptic vesicle glycoprotein 2B; synaptic vesicle protein 2B homolog
Gene Aliases: HsT19680; KIAA0735; SV2B
UniProt ID: (Human) Q7L1I2
Entrez Gene ID: (Human) 9899
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support