Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Reconstitution: 1 X 50 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
The absorption of vitamin C into the body and its distribution to organs requires two sodium-dependent vitamin C transporters. This gene encodes one of the two required transporters and the encoded protein accounts for tissue-specific uptake of vitamin C. Previously, this gene had an official symbol of SLC23A1.The absorption of vitamin C into the body and its distribution to organs requires two sodium-dependent vitamin C transporters. This gene encodes one of the two required transporters and the encoded protein accounts for tissue-specific uptake of vitamin C. Previously, this gene had an official symbol of SLC23A1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hSVCT2; mSVCT2; Na(+)/L-ascorbic acid transporter 2; Nucleobase transporter-like 1 protein; sodium-coupled ascorbic acid transporter 2; Sodium-dependent vitamin C transporter 2; sodium-dependent vitamin C transporter-2; solute carrier family 23 (ascorbic acid transporter), member 2; solute carrier family 23 (nucleobase transporters), member 1; solute carrier family 23 (nucleobase transporters), member 2; Solute carrier family 23 member 2; SVCT-2; testicular secretory protein Li 48; Yolk sac permease-like molecule 2
Gene Aliases: AI844736; hSVCT2; KIAA0238; mKIAA0238; NBTL1; SLC23A1; SLC23A2; SVCT2; YSPL2; YSPL3
UniProt ID: (Human) Q9UGH3, (Mouse) Q9EPR4, (Rat) Q9WTW8
Entrez Gene ID: (Human) 9962, (Mouse) 54338, (Rat) 50622
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support