Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SYAP1 plays a role in adipocyte differentiation by promoting mTORC2-mediated phosphorylation of AKT1 at 'Ser-473' after growth factor stimulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BSD domain-containing signal transducer and Akt interactor protein; BSTA; SAP47 homolog; synapse associated protein 1, SAP47 homolog; Synapse-associated protein 1
Gene Aliases: 2010110O17Rik; AW011796; PRO3113; SYAP1
UniProt ID: (Human) Q96A49, (Mouse) Q9D5V6
Entrez Gene ID: (Chicken) 418619, (Human) 94056, (Mouse) 67043, (Rat) 302678
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support