Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 16
Immunogen sequence: RKLLTLEDK ELGSGNFGTV KKGYYQMKKV VKTVAVKILK NEANDPALKD ELLAEANVMQ QLDNPYIVRM IGICEAESWM LVMEMAELGP LNKYLQQNRH VKDKNIIELV HQVSMGMKYL EESNFVHRDL AARNVLLVTQ HYAKISDFGL SKALRADENY YKAQTHGKWP VKWYAPECIN YYKFSSKSDV WSFGVLMWEA FSYGQKPYRG MKGSEVTAML EKGERMGCPA GCPREMYDLM NLCWTYDVEN RPGFAAVELR LRNYYYDVVN (367-635 aa encoded by BC001645)
Spleen tyrosine kinase (SYK), along with ZAP70, are cytoplasmic tyrosine kinases that are viewed as potential druge targets for allergic disorders and autoimmune diseases. SYK may also play a role in cancer. Syk translocates to the plasma membrane upon B cell antigen receptor (BCR) or the high-affinity IgE receptor (FcepsilonRI) triggering, and phosphorylates downstream adaptor proteins, thereby providing docking sites for initiation of subsequent signaling pathways, such as calcium mobilization, cytoskeleton remodeling, or transcription of specific genes. Syk binds to the receptor assemblies through interactions of its pair of SH2 domains with ITAM motives of the receptor, which have been phosphorylated by Src-family kinases. These kinases also help to activate Syk by phosphorylation of its activation loop.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp313N1010; EC 2.7.10.2; FLJ25043; FLJ37489; HGNC:11205; kinase Syk; KSYK; p72-Syk; p72Syk; SP1; Spleen tyrosine kinase; syk signaling; tyrosine-kinase syk; Tyrosine-protein kinase SYK
Gene Aliases: p72-Syk; p72syk; ptk72; SYK; Sykb
UniProt ID: (Human) P43405, (Rat) Q64725, (Mouse) P48025
Entrez Gene ID: (Human) 6850, (Rat) 25155, (Mouse) 20963
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support