Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PVVALIDID IFEVEAEERQ NIYKEVIAVQ GPPDGTVLVS IKSSLPENNF FDDALIDELL QQFASFGEVI LIRFVEDKMW VTFLEGSSAL NVLSLNGKEL LNRTITIALK SPDWIKNLEE EMSLEKISIA LPSSTSSTLL GEDAEVAADF DMEGDVDDYS AEVEELLPQH LQPSSSSGLG TSPSSSPRTS PCQSPTISEG PVPSLPIRPS RAPSRTPGPP SAQSSPIDAQ PATPLPQKDP AQPLEPKRPP PPRPVAPPTR PAPPQRPPPP S (856-1125 aa encoded by BC136603)
Synaptojanin 1 is a phosphoinositide phosphatase that regulates levels of membrane phosphatidylinositol-4,5-bisphosphate. As such, expression of this enzyme may affect synaptic transmission and membrane trafficking.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: inositol 5'-phosphatase (synaptojanin 1); inositol polyphosphate-5-phosphatase G; phosphoinositide 5-phosphatase; Synaptic inositol 1,4,5-trisphosphate 5-phosphatase 1; synaptic inositol-1; synaptic inositol-1,4,5-trisphosphate 5-phosphatase 1; Synaptojanin-1; synaptojanin-1, polyphosphoinositide phosphatase
Gene Aliases: A930006D20Rik; AA675315; INPP5G; KIAA0910; mKIAA0910; PARK20; SYNJ1
UniProt ID: (Human) O43426
Entrez Gene ID: (Human) 8867, (Mouse) 104015
Molecular Function:
phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support