Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This antibody works well in WB, IHC and IF applications.
Immunogen sequence: IEFGVLSAK KEPIQPSVLR RTYNPDDYFR KFEPHLYSLD SNSDDVDSLT DEEILSKYQL GMLHFSTQYD LLHNHLTVRV IEARDLPPPI SHDGSRQDMA HSNPYVKICL LPDQKNSKQT GVKRKTQKPV FEERYTFEIP FLEAQRRTLL LTVVDFDKFS RHCVIGKVSV PLCEVDLVKG GHWWKALIPS SQNEVELGEL LLSLNYLPSA GRLNVDVIRA KQLLQTDVSQ GSDPFVKIQL VHGLKLVKTK KTSFLRGTID PFYNESFSFK VPQEELENAS LVFTVFGHNM KSSNDFIGRI VIGQYSSGPS ETNHWRRMLN THRTAVEQWH SLRSRAECDR VSPASLEVT (127-474 aa encoded by BC004518)
Plays a role in dendrite formation by melanocytes. Classified as a brain and kidney protein (not to be confused with the BigK K+ channel); may play a role in cAMP mediated signaling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B/K protein; brain and kidney protein; Protein B/K; Synaptotagmin XVII; Synaptotagmin-17; SytXVII
Gene Aliases: Bk; SYT17; sytXVII
UniProt ID: (Human) Q9BSW7, (Rat) Q62807, (Mouse) Q920M7
Entrez Gene ID: (Human) 51760, (Rat) 192189, (Mouse) 110058
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support