Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SCGTLGFLV AAEIRIIPAK KYVKLRFEPV RGLEAICAKF THESQRQENH FVEGLLYSLD EAVIMTGVMT DEAEPSKLNS IGNYYKPWFF KHVENYLKTN REGLEYIPLR HYYHRHTRSI FWELQDIIPF GNNPIFRYLF GWMVPPKISL LKLTQGETLR KLYEQHHVVQ DMLVPMKCLQ QALHTFQNDI HVYPIWLCPF ILPSQPGLVH PKGNEAELYI DIGAYGEPRV KHFEARSCMR QLEKFVRSVH GFQMLYADCY MNREEFWEMF DGSLYHKLRE KLGCQDAFPE VYDKICKAAR H (217-516 aa encoded by BC004375)
DHCR24 is a flavin adenine dinucleotide (FAD)-dependent oxidoreductase which catalyzes the reduction of the delta-24 double bond of sterol intermediates during cholesterol biosynthesis. This protein contains a leader sequence that directs it to the endoplasmic reticulum membrane. Missense mutations in this gene have been associated with desmosterolosis. Also, reduced expression of its gene occurs in the temporal cortex of Alzheimer disease patients and overexpression has been observed in adrenal gland cancer cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 24-dehydrocholesterol reductase; 3 beta-hydroxysterol delta 24-reductase; 3-beta-hydroxysterol delta-24 reductase; 3-beta-hydroxysterol Delta-24-reductase; Delta(24)-sterol reductase; desmosterol-to-cholesterol enzyme; Diminuto/dwarf1 homolog; seladin 1; Seladin-1; selective AD indicator 1
Gene Aliases: 2310076D10Rik; 5830417J06Rik; DCE; DHCR24; KIAA0018; mKIAA0018; Nbla03646; seladin-1; SELADIN1
UniProt ID: (Human) Q15392, (Mouse) Q8VCH6
Entrez Gene ID: (Human) 1718, (Mouse) 74754
Molecular Function:
reductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support