Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
FIGURE: 1 / 3
Reconstitution: 1 X 50 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
CD108 (Sema7A) is a GPI-anchored semaphorin family member, which enhances central and peripheral axonal growth and is required for proper axon track formation during ebryogenesis. CD108 also regulates osteoclast differentiation and pre-osteoblastic cell migration, and in immune system affects cell proliferation, chemotaxis and cytokine release. On erythrocytes CD108 defines the JMH (John-Milton-Hagen) human blood group. CD108 signalizes through its receptors - plexin C1 and beta1 integrins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD108; CDw108; H-Sema K1; JMH blood group antigen; John Milton Hagen blood group H-Sema K1; John-Milton-Hargen human blood group Ag; MGC126692; MGC126696; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A (JMH blood group); sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7A; Sema K1; Sema L; semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group); Semaphorin K1; Semaphorin-7A; Semaphorin-K1; Semaphorin-L
Gene Aliases: 2900057C09Rik; CD108; CDw108; H-SEMA-K1; H-Sema-L; JMH; M-Sema-L; SEMA7A; SEMAK1; SEMAL; Semk1
UniProt ID: (Human) O75326, (Mouse) Q9QUR8
Entrez Gene ID: (Human) 8482, (Mouse) 20361, (Rat) 315711
Molecular Function: membrane-bound signaling molecule intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support