Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GSRGYIPIYTP FFMRKEVMQE VAQLSQFDEE LYKVIGKGSE KSDDNSYDEK YLIATSEQPI AALHRDEWLR PEDLPIKYAG LSTCFRQ
Highest antigen sequence identity to the following orthologs - mouse 99%, rat 97%.
Seryl-tRNA synthetase (SerRS) catalyzes the attachment of serine to tRNA(Ser) in a two-step reaction: serine is first activated by ATP to form Ser-AMP and then transferred to the acceptor end of tRNA(Ser). Is also probably able to aminoacylate tRNA(Sec) with serine, to form the misacylated tRNA L-seryl-tRNA(Sec), which will be further converted into selenocysteinyl-tRNA(Sec).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 6.1.1.11; serine tRNA ligase 1, cytoplasmic; Serine--tRNA ligase, cytoplasmic; SerRS; Seryl-tRNA synthetase; seryl-tRNA synthetase, cytoplasmic; Seryl-tRNA(Ser/Sec) synthetase
Gene Aliases: SARS; SARS1; SERRS; SERS
UniProt ID: (Human) P49591
Entrez Gene ID: (Human) 6301
Molecular Function:
aminoacyl-tRNA synthetase
translational protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support