Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Required for CNS development; midline glial cells. Involved in the metabolism of insect hormones; responsible for all ecdysone 20-monooxygenase activity during embryonic, larval and adult stages. May be involved in the breakdown of synthetic insecticides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AT11204p; CG13478-PB; CG13478-PC; CG13478-PD; CYPCCCXIVA1; cytochrome P450; Cytochrome P450 314a1, mitochondrial; E20MO; Ecdysone 20-monooxygenase; Protein shade; shd-PB; shd-PC; shd-PD
Gene Aliases: 314a1; CG13478; CYP314A1; Dmel\CG13478; Dmel_CG13478; shd
UniProt ID: (Fruit fly) Q9VUF8
Entrez Gene ID: (Fruit fly) 39592
Molecular Function: oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support