Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Major scaffold postsynaptic density protein which interacts with multiple proteins and complexes to orchestrate the dendritic spine and synapse formation, maturation and maintenance. Interconnects receptors of the postsynaptic membrane including NMDA-type and metabotropic glutamate receptors via complexes with GKAP/PSD-95 and HOMER, respectively, and the actin-based cytoskeleton. Plays a role in the structural and functional organization of the dendritic spine and synaptic junction through the interaction with Arp2/3 and WAVE1 complex as well as the promotion of the F-actin clusters. By way of this control of actin dynamics, participates in the regulation of developing neurons growth cone motility and the NMDA receptor-signaling. Also modulates GRIA1 exocytosis and GRM5/MGLUR5 expression and signaling to control the AMPA and metabotropic glutamate receptor-mediated synaptic transmission and plasticity. May be required at an early stage of synapse formation and be inhibited by IGF1 to promote synapse maturation. {PubMed:21606927, PubMed:21795692, PubMed:23100419, PubMed:23897824, PubMed:24089484}.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GKAP/SAPAP interacting protein; GKAP/SAPAP-interacting protein; proline rich synapse associated protein 2; Proline-rich synapse-associated protein 2; ProSAP2; SAPAP-interacting protein synamon; SH3 and multiple ankyrin repeat domains 1; SH3 and multiple ankyrin repeat domains protein 1; SH3 and multiple ankyrin repeat domains protein 3; SH3 and multiple ankyrin repeat domains protein 3 variant c-3; SH3 and multiple ankyrin repeat domains protein 3 variant c-4; SH3 and multiple ankyrin repeat domains-like protein 3 variant 1; SH3 and multiple ankyrin repeat domains-like protein 3 variant 2; SH3 and multiple ankyrin repeat domains-like protein 3 variant 3; SH3 and multiple ankyrin repeat domains-like protein 3 variant c-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant c-2; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-2; SH3 and multiple ankyrin repeat domains-like protein 3 variant d-3; SH3 and multiple ankyrin repeat domains-like protein 3 variant e-1; SH3 and multiple ankyrin repeat domains-like protein 3 variant e-2; SH3/ankyrin domain gene 1; SH3/ankyrin domain gene 3; Shank postsynaptic density protein 3a; Shank1; Shank3; shank3 splice variant II; shank3 splice variant III; shank3 splice variant IV; shank3e splice variant III; shank3e splice variant IV; Somatostatin receptor-interacting protein; SPANK-1; SPANK-2; SSTR-interacting protein; Synamon
Gene Aliases: AI841104; Kiaa1650; Prosap2; Shank1; Shank1a; Shank3; Shank3b; SHANK3c-3; SHANK3c-4; Spank1; Sstrip
UniProt ID: (Rat) Q9WV48, (Mouse) D3YZU1, (Mouse) Q4ACU6, (Rat) Q9JLU4
Entrez Gene ID: (Rat) 78957, (Mouse) 243961, (Mouse) 58234, (Rat) 59312
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support