Invitrogen
FIGURE: 1 / 1
This antibody recognizes and binds an epitope located at the C terminus of beta subunit of the Escherichia coli RNA polymerase. It specifically binds a 13-amino-acid long sequence 'SLAELLNAGLGGS'.
Peptide tag for mammalian expression (SLAELLNAGLGGS)