Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Fabgennix
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: putative phosphate-phosphoenolpyruvate translocator; Solute carrier family 35 member E4; solute carrier family 35, member E4
Gene Aliases: A330108F03Rik; AI414849; AI536442; SLC35E4
UniProt ID: (Human) Q6ICL7, (Rat) Q5RKL7, (Mouse) Q8K3D6
Entrez Gene ID: (Human) 339665, (Rat) 266687, (Mouse) 103710
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support