Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: SORLA.
This gene encodes a mosaic protein that belongs to at least two families: the vacuolar protein sorting 10 domain-containing receptor family, and the low density lipoprotein receptor family. The encoded protein also contains fibronectin type III repeats and an epidermal growth factor repeat. The encoded protein is translated as a preproprotein and likely plays roles in endocytosis and sorting. There may be an association between expression of this locus and Alzheimer's Disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ21930; FLJ39258; Gp250; LDLR relative with 11 ligand-binding repeats; Low-density lipoprotein receptor relative with 11 ligand-binding repeats; mSorLA; SorLA-1; Sortilin-related receptor; Sorting protein-related receptor containing LDLR class A repeats
Gene Aliases: 2900010L19Rik; AI596264; AW261561; gp250; LR11; mSorLA; Sorl1; SorLA
UniProt ID: (Mouse) O88307
Entrez Gene ID: (Mouse) 20660
Molecular Function: apolipoprotein membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support