Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EPNAVGIVIA HGSVGDAISV MVPDVYISDD GGYSWTKMLE GPHYYTILDS GGIIVAIEHS SRPINVIKFS TDEGQCWQTY TFTRDPIYFT GLASEPGARS MNISIWGFTE SFLTSQWVSY TIDFKDILER NCEEKDYTIW LAHSTDPEDY EDGCILGYKE QFLRLRKSSV CQNGRDYVVT KQPSICLCSL EDFLCDFGYY RPENDSKCVE QPELKGHDLE FCLYGREEHL TTNGYRKIPG DKCQGGVNPV REVKDLKKKC TSNFLSPEKQ NSKSNSVPII LAIVGLMLVT VVAGVLIVKK YVCGGRFLVH RYSVLQQHAE ANGVDGVDAL DTASHTNKSG YHDDSDEDLL E
Sortilin functions as a sorting receptor in the Golgi compartment and as a clearance receptor on the cell surface. Sortilin is required for protein transport from the Golgi apparatus to the lysosomes by a pathway that is independent of the mannose-6-phosphate receptor (M6PR). It is also required for protein transport from the Golgi apparatus to the endosomes. Sortilin promotes neuronal apoptosis by mediating endocytosis of the proapoptotic precursor forms of BDNF (proBDNF) and NGFB (proNGFB). Also, Sortilin acts as a receptor for neurotensin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 100 kDa NT receptor; Glycoprotein 110; Glycoprotein 95; Gp110; Gp95; mNTR3; Neurotensin receptor 3; NT3; NTR3; Sortilin
Gene Aliases: 2900053A11Rik; AI852375; Gp95; LDLCQ6; NT3; NTR3; Nts3; Ntsr3; SORT1
UniProt ID: (Human) Q99523, (Mouse) Q6PHU5, (Rat) O54861
Entrez Gene ID: (Human) 6272, (Mouse) 20661, (Rat) 83576
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support