Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Spectrin is an actin crosslinking and molecular scaffold protein that links the plasma membrane to the actin cytoskeleton, and functions in the determination of cell shape, arrangement of transmembrane proteins, and organization of organelles. It is a tetramer made up of alpha-beta dimers linked in a head-to-head arrangement. This gene is one member of a family of alpha-spectrin genes. The encoded protein is primarily composed of 22 spectrin repeats which are involved in dimer formation. It forms weaker tetramer interactions than non-erythrocytic alpha spectrin, which may increase the plasma membrane elasticity and deformability of red blood cells. Mutations in this gene result in a variety of hereditary red blood cell disorders, including elliptocytosis type 2, pyropoikilocytosis, and spherocytic hemolytic anemia. [provided by RefSeq, Jul 2008]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-I spectrin; alpha-spectrin 1, erythroid; elliptocytosis 2; Erythroid alpha-spectrin; erythroid spectrin alpha; hemolytic anemia; mutant alpha spectrin; neuroscience mutagenesis facility, 4; spectrin alpha 1; spectrin alpha chain, erythrocyte; Spectrin alpha chain, erythrocytic 1; spectrin, alpha, erythrocytic 1 (elliptocytosis 2); spherocytosis
Gene Aliases: AF093576; AI451697; EL2; ha; HPP; HS3; ihj; nmf4; sph; SPH3; Spna-1; Spna1; SPTA; SPTA1
UniProt ID: (Human) P02549, (Mouse) P08032
Entrez Gene ID: (Human) 6708, (Rat) 289257, (Mouse) 20739
Molecular Function:
actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support