Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SPTB (Spectrin Beta, Erythrocytic) is a Protein Coding gene. Diseases associated with SPTB include Spherocytosis, Type 2 and Elliptocytosis 3. Among its related pathways are Cytokine Signaling in Immune system and RET signaling. Gene Ontology (GO) annotations related to this gene include protein heterodimerization activity and actin filament binding. An important paralog of this gene is SPTBN1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-I spectrin; beta-spectrin 1; brain erythroid spectrin (235E); erythroid spectrin beta; HSPTB1; membrane cytoskeletal protein; Sp beta; spectrin beta 1; spectrin beta chain, erythrocyte; Spectrin beta chain, erythrocytic; Spectrin Beta Erythrocytic; spectrin beta Tandil; Spectrin Beta, Erythrocytic; spectrin R; SPTB1
Gene Aliases: AI842465; D330027P03Rik; EL3; Gm1301; HS2; HSPTB1; ja; jaundiced; mKIAA4219; SPH2; Spnb-1; Spnb1; SPTB; SPTB1
UniProt ID: (Human) P11277
Entrez Gene ID: (Human) 6710, (Rat) 314251, (Mouse) 20741
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support