Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAARHSTL DFMLGAKADG ETILKGLQSI FQEQGMAESV HTWQDHGYLA TYTNKNGSFA NLRIYPHGLV LLDLQSYDGD AQGKEEIDSI LNKVEERMKE LSQDSTGRVK RLPPIVRGGA IDRYWPTADG RLVEYDIDEV VYDEDSPYQN IKILHSKQFG NILILSGDVN LAESDLAYTR AIMGSGKEDY TGKDVLILGG GDGGILCEIV KLKPKMVTMV EIDQMVIDGC KKYMRKTCGD VLDNLKGDCY QVLIEDCIPV LKRYAKEGRE FDYVINDLTA VPISTSPEED STWEFLRLIL DLSMKVLKQD GKYFTQGNCV NLTEALSLYE EQLGRLYCPV EFSKEIVCVP SYLELWVFYT VWKKAKP (1-366 aa encoded by BC009898)
Catalyzes the production of spermine from spermidine and decarboxylated S-adenosylmethionine (dcSAM).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Spermidine aminopropyltransferase; Spermine synthase; SPMSY
Gene Aliases: AI427066; Gy; gyro; MRSR; SMS; SpmST; SPMSY; SpS; SRS
UniProt ID: (Human) P52788, (Mouse) P97355
Entrez Gene ID: (Human) 6611, (Mouse) 20603, (Rat) 363469
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support