Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Has an anti-hypocalcemic action on calcium and phosphate homeostasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Stanniocalcin-2; Stanniocalcin-related protein; STC-2; STC-related protein
Gene Aliases: AW125853; mustc2; STC-2; STC2; Stc2l; STCRP
UniProt ID: (Human) O76061, (Mouse) O88452
Entrez Gene ID: (Human) 8614, (Mouse) 20856
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support