Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Binds calmodulin in a calcium dependent manner. May function as scaffolding or signaling protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: protein phosphatase 2 regulatory subunit B'''gamma; striatin, calmodulin binding protein 4; Striatin-4; Zinedin
Gene Aliases: C80611; PPP2R6C; STRN4; ZIN; zinedin
UniProt ID: (Human) Q9NRL3, (Mouse) P58404
Entrez Gene ID: (Human) 29888, (Mouse) 97387
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support