Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitution: 1 X 50 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
This gene encodes a protein that belongs to the interferon-induced transmembrane family of proteins. A similar protein in rat is thought to regulate the development of excitatory synapses. [provided by RefSeq, Jul 2013].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dispanin subfamily C member 2; DSPC2; FLJ14220; interferon induced transmembrane protein domain containing 5; synapse differentiation induced gene 1; Synapse differentiation-inducing gene protein 1; SynDIG1; Transmembrane protein 90B
Gene Aliases: AW123113; C20orf39; DSPC2; Gm14134; H2C20orf39; IFITMD5; OTTMUSG00000015750; RGD1310753; SYNDIG1; TMEM90B
UniProt ID: (Human) Q9H7V2, (Mouse) A2ANU3, (Rat) Q58DZ9
Entrez Gene ID: (Human) 79953, (Mouse) 433485, (Rat) 362235
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support