Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: APQQPPKAR TGISKPYNVK QIKTTNAQEA EAAIRCLLEA RGGASEEALS AVAPRDLEAS SEPEPTPGAA KPETPQAPPL LPRRPPPRVP AIKKPTLRRT GKPLSPEEQF EQQTVHFTIG PPETSVEAPP VVTAPRVPPV PKPRTFQPGK AAERPSHRKP ASDEAPPGAG ASVPPPLEAP PLVPKVPPRR KKSAPAAFHL QVLQSNSQLL QGLTYNSSDS PSGHPPAAGT VFPQGDFLST SSATSPDSDG TKAMKPEAAP LLGDYQDPFW NLLHHPKLLN NTWLSKSSDP LDSGTRSPKR DPIDPVSAGA SAAKAELPPD HGHKTLGHWV TISDQEKRTA LQVFDPLAKT (1147-1496 aa encoded by BC043277)
The gene is a member of the inositol-polyphosphate 5-phosphatase family. The encoded protein interacts with the ras-related C3 botulinum toxin substrate 1, which causes translocation of the encoded protein to the plasma membrane where it inhibits clathrin-mediated endocytosis. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: inositol phosphate 5'-phosphatase 2; inositol polyphosphate-5-phosphatase H; phosphoinositide 5-phosphatase; Synaptic inositol 1,4,5-trisphosphate 5-phosphatase 2; synaptic inositol-1,4,5-trisphosphate 5-phosphatase 2; Synaptojanin-2
Gene Aliases: AI481647; INPP5H; KIAA0348; mKIAA0348; SJ2; SYNJ2
UniProt ID: (Human) O15056, (Mouse) Q9D2G5, (Rat) O55207
Entrez Gene ID: (Human) 8871, (Mouse) 20975, (Rat) 84018
Molecular Function:
phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support