Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
SYT14 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Synaptotagmin XIV; Synaptotagmin-14; SytXIV
Gene Aliases: B230320I09Rik; SCAR11; SYT14; sytXIV
UniProt ID: (Human) Q8NB59, (Mouse) Q7TN84
Entrez Gene ID: (Human) 255928, (Rat) 684293, (Mouse) 329324
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support