Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: SYTII.
Synaptotagmins, like SYT2, are integral membrane proteins of synaptic vesicles thought to serve as Ca(2+) sensors in the process of vesicular trafficking and exocytosis (Hilbush and Morgan, 1994 [PubMed 8058779]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inositol polyphosphate-binding protein; IP4-binding protein; synaptotagmin 2; Synaptotagmin II; Synaptotagmin-2; SytII
Gene Aliases: CMS7; mKIAA4194; MYSPC; R74640; RATSYNII; SYNII; SYT2; SytII
UniProt ID: (Human) Q8N9I0, (Rat) P29101, (Mouse) P46097
Entrez Gene ID: (Human) 127833, (Rat) 24805, (Mouse) 20980
Molecular Function:
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support