Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MPWRNKEASS PSSANPPLEA LQSPSFRGNM ADKLKDPSTL GFLEAAVKIS HTSPDIPAEV QMSVKEHIMR HTRLQRQTTE PASSTRHTSF KRHLPRQMHV SSVDYGNELP PAAEQPTSIG
Synaptotagmins, such as SYT6, share a common domain structure that includes a transmembrane domain and a cytoplasmic region composed of 2 C2 domains. Some synaptotagmins are involved in synaptic membrane fusion, while others have a more general function in endocytosis. For further information on synaptotagmins, see MIM 185605.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: synaptotagmin 6; Synaptotagmin VI; Synaptotagmin-6; Synaptotagmin6; SynaptotagminVI; Syt VI; SYT-6; SytVI
Gene Aliases: 3110037A08Rik; AW048930; SYT6; sytVI
UniProt ID: (Human) Q5T7P8, (Mouse) Q9R0N8, (Rat) Q62746
Entrez Gene ID: (Human) 148281, (Mouse) 54524, (Rat) 60565
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support