Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody is predicted to react with human, rat, pig and cow based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
SYT9 may be involved in Ca2+-dependent exocytosis of secretory vesicles through Ca2+ and phospholipid binding to the C2 domain or may serve as Ca2+ sensors in the process of vesicular trafficking and exocytosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ45896; synaptotagmin 9; Synaptotagmin IX; Synaptotagmin V; Synaptotagmin-9; Synaptotagmin9; SynaptotagminIX; Syt IX; SYT-9; SytIX
Gene Aliases: Syt5; Syt9
UniProt ID: (Mouse) Q9R0N9
Entrez Gene ID: (Mouse) 60510
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support